Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTTCCATGGTTGCTGTTGAGAA[A/C]ACTGATTCTATTCTGATCCTTGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604831 MIM: 607261 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EVC - EvC ciliary complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306090.1 | Intron | NP_001293019.1 | ||||
NM_001306092.1 | Intron | NP_001293021.1 | ||||
NM_153717.2 | Intron | NP_714928.1 | ||||
XM_006713865.3 | Intron | XP_006713928.1 | ||||
XM_006713866.3 | Intron | XP_006713929.1 | ||||
XM_011513419.2 | Intron | XP_011511721.1 | ||||
XM_017007883.1 | Intron | XP_016863372.1 |
EVC2 - EvC ciliary complex subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |