Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCAATGGGGAGACAGAATTCTTG[C/T]CCTTGGGATGCTGGCCTCCAGCCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172411 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G2A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLA2G2A - phospholipase A2 group IIA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000300.3 | Intron | NP_000291.1 | ||||
NM_001161727.1 | Intron | NP_001155199.1 | ||||
NM_001161728.1 | Intron | NP_001155200.1 | ||||
NM_001161729.1 | Intron | NP_001155201.1 |
Set Membership: |
HapMap |