Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAAGGTTAGAACAACTGACCTA[G/T]TGAAAGTCTTGAAGGGAAATTATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608317 | ||||||||||||||||||||
Literature Links: |
GRHL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRHL3 - grainyhead like transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195010.1 | Intron | NP_001181939.1 | ||||
NM_021180.3 | Intron | NP_067003.2 | ||||
NM_198173.2 | Intron | NP_937816.1 | ||||
NM_198174.2 | Intron | NP_937817.3 | ||||
XM_011541869.1 | Intron | XP_011540171.1 | ||||
XM_011541870.2 | Intron | XP_011540172.1 |