Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAGATTTGTATAATTCATATTG[C/T]TTTACATGGAGTCTTCCAATAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615580 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF528 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF528 - zinc finger protein 528 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032423.2 | 2848 | Intron | NP_115799.2 | |||
XM_005259323.1 | 2848 | Intron | XP_005259380.1 | |||
XM_006723418.2 | 2848 | Intron | XP_006723481.1 | |||
XM_011527392.1 | 2848 | Intron | XP_011525694.1 | |||
XM_011527394.2 | 2848 | Intron | XP_011525696.1 | |||
XM_017027360.1 | 2848 | Intron | XP_016882849.1 | |||
XM_017027361.1 | 2848 | Intron | XP_016882850.1 | |||
XM_017027362.1 | 2848 | Intron | XP_016882851.1 | |||
XM_017027363.1 | 2848 | Intron | XP_016882852.1 | |||
XM_017027364.1 | 2848 | Intron | XP_016882853.1 | |||
XM_017027365.1 | 2848 | Intron | XP_016882854.1 |
ZNF528-AS1 - ZNF528 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF880 - zinc finger protein 880 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145434.1 | 2848 | Intron | NP_001138906.1 | |||
XM_017026812.1 | 2848 | UTR 3 | XP_016882301.1 |
Set Membership: |
HapMap |