Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCTCCTTGGTTTCCTGCCTTGG[A/G]CCTTCGCTTCAGCTGTGCCCTCTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611191 MIM: 614510 MIM: 611250 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR125A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR125A - microRNA 125a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR99B - microRNA 99b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIRLET7E - microRNA let-7e | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPACA6 - sperm acrosome associated 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316972.1 | 71 | Intron | NP_001303901.1 | |||
NM_001316994.1 | 71 | Intron | NP_001303923.1 | |||
XM_017026299.1 | 71 | Intron | XP_016881788.1 | |||
XM_017026300.1 | 71 | UTR 5 | XP_016881789.1 | |||
XM_017026301.1 | 71 | Intron | XP_016881790.1 |
SPACA6P-AS - SPACA6P antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |