Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATTAACCAAAATTAACCATACA[C/T]TGGGTCACAAACCAAGTTTCAGCAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 165260 | |||||||||||||||||||||||
Literature Links: |
MIR6806 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MIR6806 - microRNA 6806 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN22 - zinc finger and SCAN domain containing 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321116.1 | Intron | NP_001308045.1 | ||||
NM_001321117.1 | Intron | NP_001308046.1 | ||||
NM_181846.2 | Intron | NP_862829.1 | ||||
XM_006723192.3 | Intron | XP_006723255.1 | ||||
XM_011526917.2 | Intron | XP_011525219.1 | ||||
XM_011526918.2 | Intron | XP_011525220.1 | ||||
XM_017026746.1 | Intron | XP_016882235.1 | ||||
XM_017026747.1 | Intron | XP_016882236.1 |
Set Membership: |
HapMap |