Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYTL5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SYTL5 - synaptotagmin like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163334.1 | Intron | NP_001156806.1 | ||||
NM_001163335.1 | Intron | NP_001156807.1 | ||||
NM_138780.2 | Intron | NP_620135.1 | ||||
XM_011544001.2 | Intron | XP_011542303.1 | ||||
XM_011544002.2 | Intron | XP_011542304.1 | ||||
XM_017029972.1 | Intron | XP_016885461.1 | ||||
XM_017029973.1 | Intron | XP_016885462.1 | ||||
XM_017029974.1 | Intron | XP_016885463.1 |