Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAGTTCACCTGGGAGTTTTAGG[A/G]AACCTGAAGACGGATGAGGGTGCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 311790 MIM: 300132 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PFKFB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PFKFB1 - 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271804.1 | Intron | NP_001258733.1 | ||||
NM_001271805.1 | Intron | NP_001258734.1 | ||||
NM_002625.3 | Intron | NP_002616.2 | ||||
XM_017029576.1 | Intron | XP_016885065.1 | ||||
XM_017029577.1 | Intron | XP_016885066.1 | ||||
XM_017029578.1 | Intron | XP_016885067.1 |
TRO - trophinin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |