Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606277 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOLLIP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TOLLIP - toll interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318512.1 | 2087 | UTR 3 | NP_001305441.1 | |||
NM_001318514.1 | 2087 | UTR 3 | NP_001305443.1 | |||
NM_001318515.1 | 2087 | UTR 3 | NP_001305444.1 | |||
NM_001318516.1 | 2087 | UTR 3 | NP_001305445.1 | |||
NM_019009.3 | 2087 | UTR 3 | NP_061882.2 | |||
XM_017017931.1 | 2087 | UTR 3 | XP_016873420.1 |