Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCCGAGAGGTTGGAGTTGGGGC[A/G]ATGGGGCTGAGAGAGGTCGGAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603870 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBFA2T3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CBFA2T3 - CBFA2/RUNX1 translocation partner 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005187.5 | 3299 | UTR 3 | NP_005178.4 | |||
NM_175931.2 | 3299 | UTR 3 | NP_787127.1 | |||
XM_005256323.4 | 3299 | UTR 3 | XP_005256380.1 | |||
XM_011523419.2 | 3299 | Intron | XP_011521721.1 | |||
XM_017023811.1 | 3299 | Intron | XP_016879300.1 |
LOC101927793 - uncharacterized LOC101927793 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PABPN1L - poly(A) binding protein nuclear 1 like (cytoplasmic) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |