Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTCAGACTTTACATTAGAAAC[C/T]CATTAAAGCCTACTTCTTTCTTGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300401 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLP1 - proteolipid protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000533.4 | Intron | NP_000524.3 | ||||
NM_001128834.2 | Intron | NP_001122306.1 | ||||
NM_001305004.1 | Intron | NP_001291933.1 | ||||
NM_199478.2 | Intron | NP_955772.1 |
Set Membership: |
HapMap |