Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAGCTGTTTATCAGGGACTGTC[A/G]AATCCAACAGCTGTTTACACAGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194537 | ||||||||||||||||||||||||||||||||
Literature Links: |
ZNF26 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ZNF26 - zinc finger protein 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256279.1 | Intron | NP_001243208.1 | ||||
NM_001256280.1 | Intron | NP_001243209.1 | ||||
NM_019591.3 | Intron | NP_062537.2 | ||||
XM_005266182.3 | Intron | XP_005266239.1 | ||||
XM_005266183.3 | Intron | XP_005266240.1 | ||||
XM_011534829.1 | Intron | XP_011533131.1 | ||||
XM_017019920.1 | Intron | XP_016875409.1 | ||||
XM_017019921.1 | Intron | XP_016875410.1 | ||||
XM_017019922.1 | Intron | XP_016875411.1 | ||||
XM_017019923.1 | Intron | XP_016875412.1 |