Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAACGGTGAAGGTGGGGGTGTG[T/C]GCGTCATTTGAAAGATGTATAGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606248 MIM: 182160 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
QPRT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
QPRT - quinolinate phosphoribosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318249.1 | Intron | NP_001305178.1 | ||||
NM_001318250.1 | Intron | NP_001305179.1 | ||||
NM_014298.4 | Intron | NP_055113.2 | ||||
XM_005255223.3 | Intron | XP_005255280.2 | ||||
XM_017023101.1 | Intron | XP_016878590.1 |
SPN - sialophorin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |