Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATCTGCCTTTTTCCTCCTGTTA[T/C]ACAGATGGCAAGACCATGGACTATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR3C PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
POLR3C - RNA polymerase III subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303456.1 | Intron | NP_001290385.1 | ||||
NM_006468.7 | Intron | NP_006459.3 | ||||
XM_005272927.3 | Intron | XP_005272984.1 | ||||
XM_005272928.3 | Intron | XP_005272985.1 | ||||
XM_011509078.2 | Intron | XP_011507380.1 |
RNF115 - ring finger protein 115 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |