Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCATAAAAATCGTGTTTTTTTT[A/T]ATATTGTGCTCACATGTTGTTTTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602597 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCL9 - B-cell CLL/lymphoma 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004326.3 | Intron | NP_004317.2 | ||||
XM_005272971.4 | Intron | XP_005273028.1 | ||||
XM_005277417.3 | Intron | XP_005277474.1 | ||||
XM_005277418.3 | Intron | XP_005277475.1 | ||||
XM_017002026.1 | Intron | XP_016857515.1 |
Set Membership: |
HapMap |