Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGCAGACCAGTGAAAGCACCCT[C/G]GTGGCGCGGGGCCCAGATGTCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613374 MIM: 171150 MIM: 601292 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGF29 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SGF29 - SAGA complex associated factor 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SULT1A1 - sulfotransferase family 1A member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SULT1A2 - sulfotransferase family 1A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001054.3 | 111 | Intron | NP_001045.1 | |||
NM_177528.2 | 111 | UTR 5 | NP_803564.1 | |||
XM_006721077.3 | 111 | UTR 5 | XP_006721140.1 | |||
XM_017023600.1 | 111 | Intron | XP_016879089.1 | |||
XM_017023601.1 | 111 | UTR 5 | XP_016879090.1 | |||
XM_017023602.1 | 111 | UTR 5 | XP_016879091.1 | |||
XM_017023603.1 | 111 | Intron | XP_016879092.1 |
Set Membership: |
DME Validated Inventoried |