Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTCCATATCCCACTGCCAAAAC[C/T]GCATGGTTCAGATTATCGCTATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
75 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601105 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTSK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CTSK - cathepsin K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000396.3 | 1060 | Silent Mutation | GCA,GCG | A,A 277 | NP_000387.1 |
Set Membership: |
HapMap |