Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCTCTGAGTGGGGAGTTGGGAA[G/T]AAATGTTCTAGCACACCACTAACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612208 MIM: 609824 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLPH3L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GOLPH3L - golgi phosphoprotein 3 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HORMAD1 - HORMA domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199829.1 | Intron | NP_001186758.1 | ||||
NM_032132.4 | Intron | NP_115508.2 | ||||
XM_006711570.2 | Intron | XP_006711633.1 | ||||
XM_006711571.2 | Intron | XP_006711634.1 | ||||
XM_011510053.2 | Intron | XP_011508355.1 | ||||
XM_011510054.2 | Intron | XP_011508356.1 | ||||
XM_011510055.2 | Intron | XP_011508357.1 | ||||
XM_011510056.2 | Intron | XP_011508358.1 |