Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAGTCACAGAAAAAAACAGGGA[A/C]CAGTCAAAAACAATAAAAGAATAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609611 | ||||||||||||||||||||
Literature Links: |
ANP32E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANP32E - acidic nuclear phosphoprotein 32 family member E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136478.3 | 3143 | UTR 3 | NP_001129950.1 | |||
NM_001136479.2 | 3143 | UTR 3 | NP_001129951.1 | |||
NM_001280559.1 | 3143 | UTR 3 | NP_001267488.1 | |||
NM_001280560.1 | 3143 | UTR 3 | NP_001267489.1 | |||
NM_030920.4 | 3143 | UTR 3 | NP_112182.1 | |||
XM_005245513.3 | 3143 | UTR 3 | XP_005245570.1 | |||
XM_005245514.3 | 3143 | UTR 3 | XP_005245571.1 | |||
XM_017002418.1 | 3143 | UTR 3 | XP_016857907.1 |