Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTCTCCCTCCTCTCTCCCCTTC[A/G]CCCACTCACTCTACCCATCCACCTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZDHHC19 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZDHHC19 - zinc finger DHHC-type containing 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039617.1 | Intron | NP_001034706.1 | ||||
XM_005269279.2 | Intron | XP_005269336.1 | ||||
XM_006713493.2 | Intron | XP_006713556.1 | ||||
XM_006713494.2 | Intron | XP_006713557.1 | ||||
XM_011512417.1 | Intron | XP_011510719.1 |