Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGTGCCACCTGTAGACATGAAGT[C/T]AAAAGATTAGACACTTGAGTACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102750 MIM: 610366 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADK - adenosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123.3 | Intron | NP_001114.2 | ||||
NM_001202449.1 | Intron | NP_001189378.1 | ||||
NM_001202450.1 | Intron | NP_001189379.1 | ||||
NM_006721.3 | Intron | NP_006712.2 | ||||
XM_017015698.1 | Intron | XP_016871187.1 | ||||
XM_017015699.1 | Intron | XP_016871188.1 | ||||
XM_017015700.1 | Intron | XP_016871189.1 | ||||
XM_017015701.1 | Intron | XP_016871190.1 | ||||
XM_017015702.1 | Intron | XP_016871191.1 | ||||
XM_017015703.1 | Intron | XP_016871192.1 | ||||
XM_017015704.1 | Intron | XP_016871193.1 | ||||
XM_017015705.1 | Intron | XP_016871194.1 | ||||
XM_017015706.1 | Intron | XP_016871195.1 |
AP3M1 - adaptor related protein complex 3 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |