Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTTTTAAGCTCTAATTGCTAA[C/T]TGGTTGGTGTCTGAAGGGACCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605880 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAT6B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KAT6B - lysine acetyltransferase 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256468.1 | Intron | NP_001243397.1 | ||||
NM_001256469.1 | Intron | NP_001243398.1 | ||||
NM_012330.3 | Intron | NP_036462.2 | ||||
XM_005269664.2 | Intron | XP_005269721.1 | ||||
XM_017016000.1 | Intron | XP_016871489.1 | ||||
XM_017016001.1 | Intron | XP_016871490.1 | ||||
XM_017016002.1 | Intron | XP_016871491.1 | ||||
XM_017016003.1 | Intron | XP_016871492.1 | ||||
XM_017016004.1 | Intron | XP_016871493.1 | ||||
XM_017016005.1 | Intron | XP_016871494.1 | ||||
XM_017016006.1 | Intron | XP_016871495.1 | ||||
XM_017016007.1 | Intron | XP_016871496.1 | ||||
XM_017016008.1 | Intron | XP_016871497.1 | ||||
XM_017016009.1 | Intron | XP_016871498.1 |
Set Membership: |
HapMap |