Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACAAAAATATTTCCTTAAAAAG[C/T]TGTTAAAAGTTTTTTAGTCCTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613133 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TSPAN2 - tetraspanin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308315.1 | 1404 | UTR 3 | NP_001295244.1 | |||
NM_001308316.1 | 1404 | UTR 3 | NP_001295245.1 | |||
NM_005725.5 | 1404 | UTR 3 | NP_005716.2 | |||
XM_016999996.1 | 1404 | UTR 3 | XP_016855485.1 |