Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCAAGGAGAGACAAAGATGGGA[A/G]AGACAGCAAGAGATGGGGGAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601855 MIM: 112205 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARHGEF1 - Rho guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004706.3 | Intron | NP_004697.2 | ||||
NM_198977.1 | Intron | NP_945328.1 | ||||
NM_199002.1 | Intron | NP_945353.1 | ||||
XM_005259386.4 | Intron | XP_005259443.1 | ||||
XM_005259390.4 | Intron | XP_005259447.1 | ||||
XM_011527468.2 | Intron | XP_011525770.1 |
CD79A - CD79a molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |