Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGGGGGCAGAAAGTCACCTCCT[G/T]CTCTCCCTCCACTGTCCACAGAGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608083 MIM: 600745 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOC2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APOC2 - apolipoprotein C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOC4 - apolipoprotein C4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001646.2 | Intron | NP_001637.1 |
APOC4-APOC2 - APOC4-APOC2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |