Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGGAGGGAATAATTGGACATCT[C/T]GTTTTCTTTCATGCTCTGAAAGGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125263 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01595 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
LINC01595 - long intergenic non-protein coding RNA 1595 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SULT2A1 - sulfotransferase family 2A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003167.3 | 819 | Missense Mutation | CAG,TAG | Q,* 227 | NP_003158.2 |
Set Membership: |
HapMap DME Validated Inventoried |