Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAAGGGTAGCGGCTGAGTCGAC[A/C]GGCCTCCATGTCCCCTCTCCCTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603604 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAIAP2L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAIAP2L2 - BAI1 associated protein 2 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G6 - phospholipase A2 group VI | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004426.1 | Intron | NP_001004426.1 | ||||
NM_001199562.1 | Intron | NP_001186491.1 | ||||
NM_003560.2 | Intron | NP_003551.2 | ||||
XM_005261764.2 | Intron | XP_005261821.1 | ||||
XM_005261765.1 | Intron | XP_005261822.1 | ||||
XM_005261766.1 | Intron | XP_005261823.1 | ||||
XM_006724332.3 | Intron | XP_006724395.1 | ||||
XM_011530422.1 | Intron | XP_011528724.1 | ||||
XM_011530423.1 | Intron | XP_011528725.1 | ||||
XM_011530424.1 | Intron | XP_011528726.1 | ||||
XM_011530425.1 | Intron | XP_011528727.1 | ||||
XM_011530426.2 | Intron | XP_011528728.1 | ||||
XM_017028981.1 | Intron | XP_016884470.1 | ||||
XM_017028982.1 | Intron | XP_016884471.1 | ||||
XM_017028983.1 | Intron | XP_016884472.1 | ||||
XM_017028984.1 | Intron | XP_016884473.1 | ||||
XM_017028985.1 | Intron | XP_016884474.1 | ||||
XM_017028986.1 | Intron | XP_016884475.1 | ||||
XM_017028987.1 | Intron | XP_016884476.1 | ||||
XM_017028988.1 | Intron | XP_016884477.1 |