Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATGGAACCATCACCATAATCC[A/C]GTTTTAGAACAGTTCTCAAAGAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606938 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UROS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UROS - uroporphyrinogen III synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000375.2 | Intron | NP_000366.1 | ||||
NM_001324036.1 | Intron | NP_001310965.1 | ||||
NM_001324037.1 | Intron | NP_001310966.1 | ||||
NM_001324038.1 | Intron | NP_001310967.1 | ||||
NM_001324039.1 | Intron | NP_001310968.1 | ||||
XM_005270140.4 | Intron | XP_005270197.1 | ||||
XM_011540127.1 | Intron | XP_011538429.1 | ||||
XM_017016610.1 | Intron | XP_016872099.1 | ||||
XM_017016611.1 | Intron | XP_016872100.1 | ||||
XM_017016612.1 | Intron | XP_016872101.1 |
Set Membership: |
HapMap |