Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACCAAGAAGTCCTTCAAGACTCT[C/T]CACTTTATTTGCCTAGATAAGACAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602856 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RGS10 - regulator of G-protein signaling 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005339.1 | Intron | NP_001005339.1 | ||||
NM_002925.3 | Intron | NP_002916.1 | ||||
XM_017016492.1 | Intron | XP_016871981.1 | ||||
XM_017016493.1 | Intron | XP_016871982.1 |