Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGTTTTGGTTATCCCTCAGTGT[C/T]AGTGTGGACCTGTCCCAGGTTGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615662 | ||||||||||||||||||||
Literature Links: |
SERPINB12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERPINB12 - serpin family B member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307928.1 | Intron | NP_001294857.1 | ||||
NM_080474.2 | Intron | NP_536722.1 | ||||
XM_005266778.3 | Intron | XP_005266835.2 | ||||
XM_011526246.1 | Intron | XP_011524548.1 | ||||
XM_011526248.1 | Intron | XP_011524550.1 | ||||
XM_011526249.1 | Intron | XP_011524551.1 | ||||
XM_017026067.1 | Intron | XP_016881556.1 |