Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTGCTGAAATGTCAAAATGTAA[A/G]ATTGAATGAGGCTATTTAAACATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZBTB44 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBTB44 - zinc finger and BTB domain containing 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301098.1 | 5515 | UTR 3 | NP_001288027.1 | |||
NM_001301099.1 | 5515 | UTR 3 | NP_001288028.1 | |||
NM_014155.4 | 5515 | UTR 3 | NP_054874.3 | |||
XM_005271521.4 | 5515 | UTR 3 | XP_005271578.1 | |||
XM_005271522.4 | 5515 | UTR 3 | XP_005271579.1 | |||
XM_006718825.3 | 5515 | UTR 3 | XP_006718888.1 | |||
XM_006718826.3 | 5515 | UTR 3 | XP_006718889.1 | |||
XM_011542789.2 | 5515 | Intron | XP_011541091.1 | |||
XM_017017623.1 | 5515 | UTR 3 | XP_016873112.1 | |||
XM_017017624.1 | 5515 | UTR 3 | XP_016873113.1 |