Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603312 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BBOX1 - gamma-butyrobetaine hydroxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003986.2 | Intron | NP_003977.1 | ||||
XM_005253159.2 | Intron | XP_005253216.1 | ||||
XM_005253160.2 | Intron | XP_005253217.1 | ||||
XM_005253161.2 | Intron | XP_005253218.1 | ||||
XM_011520402.1 | Intron | XP_011518704.1 | ||||
XM_017018403.1 | Intron | XP_016873892.1 |
BBOX1-AS1 - BBOX1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |