Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGGGAGGACCTGGTGTTGATTC[C/T]ATCAAGATATGACAAAGTTTATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
64 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLD5 - phospholipase D family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195811.1 | 3298 | UTR 3 | NP_001182740.1 | |||
NM_001195812.1 | 3298 | UTR 3 | NP_001182741.1 | |||
NM_001320272.1 | 3298 | UTR 3 | NP_001307201.1 | |||
NM_152666.2 | 3298 | UTR 3 | NP_689879.2 | |||
XM_006711752.2 | 3298 | UTR 3 | XP_006711815.1 | |||
XM_011544115.2 | 3298 | UTR 3 | XP_011542417.1 | |||
XM_011544116.2 | 3298 | UTR 3 | XP_011542418.1 | |||
XM_011544119.2 | 3298 | UTR 3 | XP_011542421.2 | |||
XM_011544120.2 | 3298 | UTR 3 | XP_011542422.1 | |||
XM_011544121.2 | 3298 | UTR 3 | XP_011542423.1 | |||
XM_011544122.2 | 3298 | UTR 3 | XP_011542424.1 | |||
XM_017000567.1 | 3298 | Intron | XP_016856056.1 | |||
XM_017000568.1 | 3298 | UTR 3 | XP_016856057.1 | |||
XM_017000569.1 | 3298 | UTR 3 | XP_016856058.1 | |||
XM_017000570.1 | 3298 | UTR 3 | XP_016856059.1 |
Set Membership: |
HapMap |