Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTGTGAAAGAATTTGTGGCTGA[A/G]AAACTGGCCGCACTCCTTTGTAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF347 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF347 - zinc finger protein 347 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF665 - zinc finger protein 665 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024733.3 | 2486 | UTR 3 | NP_079009.3 | |||
XM_005259266.4 | 2486 | Intron | XP_005259323.1 | |||
XM_011527325.2 | 2486 | Intron | XP_011525627.1 | |||
XM_011527327.2 | 2486 | Intron | XP_011525629.1 |
Set Membership: |
HapMap |