Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606427 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF320 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF320 - zinc finger protein 320 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207333.2 | Intron | NP_997216.2 | ||||
XM_006723059.3 | Intron | XP_006723122.1 | ||||
XM_017026393.1 | Intron | XP_016881882.1 | ||||
XM_017026394.1 | Intron | XP_016881883.1 | ||||
XM_017026395.1 | Intron | XP_016881884.1 | ||||
XM_017026396.1 | Intron | XP_016881885.1 | ||||
XM_017026397.1 | Intron | XP_016881886.1 | ||||
XM_017026398.1 | Intron | XP_016881887.1 | ||||
XM_017026399.1 | Intron | XP_016881888.1 | ||||
XM_017026400.1 | Intron | XP_016881889.1 |