Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607874 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF444 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF444 - zinc finger protein 444 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253792.1 | 33 | UTR 5 | NP_001240721.1 | |||
NM_018337.3 | 33 | UTR 5 | NP_060807.2 | |||
XM_005259034.2 | 33 | UTR 5 | XP_005259091.1 | |||
XM_005259035.1 | 33 | Intron | XP_005259092.1 | |||
XM_005259036.1 | 33 | Intron | XP_005259093.1 | |||
XM_011527068.1 | 33 | Intron | XP_011525370.1 | |||
XM_011527069.1 | 33 | Intron | XP_011525371.1 | |||
XM_011527070.1 | 33 | Intron | XP_011525372.1 | |||
XM_011527071.1 | 33 | Intron | XP_011525373.1 | |||
XM_011527073.1 | 33 | Intron | XP_011525375.1 | |||
XM_017026932.1 | 33 | Intron | XP_016882421.1 | |||
XM_017026933.1 | 33 | Intron | XP_016882422.1 |
Set Membership: |
HapMap |