Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCTATTTTCTTAGTCTCTGTT[C/T]TATTTGTGCTTTGATCTTTTTTATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300092 MIM: 300044 | ||||||||||||||||||||
Literature Links: |
TEX28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEX28 - testis expressed 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TKTL1 - transketolase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145933.1 | Intron | NP_001139405.1 | ||||
NM_001145934.1 | Intron | NP_001139406.1 | ||||
NM_012253.3 | Intron | NP_036385.3 |