Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGAGCAGTATTATAGATAAACA[A/G]TTAAATGCAGATGTATCCGTAATGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607813 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLPPR4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLPPR4 - phospholipid phosphatase related 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166252.1 | Intron | NP_001159724.1 | ||||
NM_014839.4 | Intron | NP_055654.2 | ||||
XM_011542498.2 | Intron | XP_011540800.1 |
Set Membership: |
HapMap |