Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAAAATTGTCAATATTCAACCAG[C/T]TATGACAAAAATGGTACTTCATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610656 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KYAT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KYAT3 - kynurenine aminotransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008661.2 | Intron | NP_001008661.1 | ||||
NM_001008662.2 | Intron | NP_001008662.1 |
RBMXL1 - RNA binding motif protein, X-linked like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162536.2 | Intron | NP_001156008.1 | ||||
NM_019610.5 | Intron | NP_062556.2 |
Set Membership: |
HapMap |