Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGAGCCCAGAAAACTTCTCTA[A/T]GGATGTAATTTTTGAGCTAAGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608057 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNASE2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNASE2B - deoxyribonuclease II beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021233.2 | Intron | NP_067056.2 | ||||
NM_058248.1 | Intron | NP_490649.1 | ||||
XM_011541878.2 | Intron | XP_011540180.1 |
Set Membership: |
HapMap |