Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACCCGCACACCCCCGCCCCGTA[C/T]AGGCGTGTCTCTCTGTGACCCGAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600874 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GNG5 - G protein subunit gamma 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005274.2 | Intron | NP_005265.1 |
RPF1 - ribosome production factor 1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA1 - spermatogenesis associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310156.1 | Intron | NP_001297085.1 | ||||
XM_011542514.2 | Intron | XP_011540816.1 | ||||
XM_011542515.2 | Intron | XP_011540817.1 | ||||
XM_011542516.1 | Intron | XP_011540818.1 | ||||
XM_011542517.1 | Intron | XP_011540819.1 | ||||
XM_011542519.2 | Intron | XP_011540821.1 | ||||
XM_011542520.2 | Intron | XP_011540822.1 | ||||
XM_011542521.2 | Intron | XP_011540823.1 | ||||
XM_011542522.2 | Intron | XP_011540824.1 |
Set Membership: |
HapMap |