Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAATAAGACACAGCAATGAAAA[C/T]AACTGCTTCTCCACTCAACAACGTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601442 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CFL1 - cofilin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX32 - sorting nexin 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152760.2 | Intron | NP_689973.2 | ||||
XM_006718488.3 | Intron | XP_006718551.1 | ||||
XM_017017470.1 | Intron | XP_016872959.1 | ||||
XM_017017471.1 | Intron | XP_016872960.1 | ||||
XM_017017472.1 | Intron | XP_016872961.1 | ||||
XM_017017473.1 | Intron | XP_016872962.1 |
Set Membership: |
HapMap |