Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGGCGGTGGCGGCGGCGGCGGC[G/T]GCGGCGGTGGTGGTGGCGGGCCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605624 | ||||||||||||||||||||
Literature Links: |
ARIH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARIH1 - ariadne RBR E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005744.3 | 10 | Missense Mutation | GGC,TGC | G,C 84 | NP_005735.2 | |
XM_011521438.2 | 10 | Silent Mutation | GCG,GCT | A,A 47 | XP_011519740.1 | |
XM_011521439.2 | 10 | UTR 5 | XP_011519741.1 |
LOC105370888 - uncharacterized LOC105370888 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |