Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTTTTCCTTCTAAGAGTATAGA[A/G]TAATGTCACTTTTAAGAATGTATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611202 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASAH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASAH2 - N-acylsphingosine amidohydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143974.1 | Intron | NP_001137446.1 | ||||
NM_019893.2 | Intron | NP_063946.2 | ||||
XM_011539970.2 | Intron | XP_011538272.1 | ||||
XM_011539971.2 | Intron | XP_011538273.1 | ||||
XM_011539972.2 | Intron | XP_011538274.1 | ||||
XM_011539973.2 | Intron | XP_011538275.1 | ||||
XM_017016435.1 | Intron | XP_016871924.1 | ||||
XM_017016436.1 | Intron | XP_016871925.1 |