Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGACGAGGAAGATGAGGATGAGGT[G/T]GAAGAGGAGGAGACCAACAGGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612757 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPIHBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPIHBP1 - glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301772.1 | 213 | Silent Mutation | GTG,GTT | V,V 46 | NP_001288701.1 | |
NM_178172.5 | 213 | Silent Mutation | GTG,GTT | V,V 46 | NP_835466.2 |