Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAGGGAGGAAGGATGCTGTAGT[A/G]TATGAAAACAAAAGTTTTCACCTGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 610345 | |||||||||||||||||||||||
Literature Links: |
AGK PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
AGK - acylglycerol kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018238.3 | 3995 | Intron | NP_060708.1 | |||
XM_005250023.3 | 3995 | Intron | XP_005250080.1 | |||
XM_011516397.2 | 3995 | UTR 3 | XP_011514699.1 |
KIAA1147 - KIAA1147 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080392.1 | 3995 | Intron | NP_001073861.1 |
Set Membership: |
HapMap |