Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACTTTCATAAACCAGACTGGGG[A/C]GAAATCTCTTTTTAAAAAAATCAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603140 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIP4K2A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PIP4K2A - phosphatidylinositol-5-phosphate 4-kinase type 2 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005028.4 | 2476 | UTR 3 | NP_005019.2 | |||
XM_006717450.2 | 2476 | UTR 3 | XP_006717513.1 | |||
XM_011519496.1 | 2476 | UTR 3 | XP_011517798.1 | |||
XM_017016330.1 | 2476 | UTR 3 | XP_016871819.1 | |||
XM_017016331.1 | 2476 | UTR 3 | XP_016871820.1 | |||
XM_017016332.1 | 2476 | UTR 3 | XP_016871821.1 |
Set Membership: |
HapMap |