Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTATACTTATGTTTTACATTT[C/T]TTCATTTATGTTTAACATTGATAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602997 MIM: 179555 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUBN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CUBN - cubilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081.3 | Intron | NP_001072.2 | ||||
XM_011519708.2 | Intron | XP_011518010.1 | ||||
XM_011519709.2 | Intron | XP_011518011.1 | ||||
XM_011519710.2 | Intron | XP_011518012.1 | ||||
XM_011519711.2 | Intron | XP_011518013.1 |
RSU1 - Ras suppressor protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |