Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACACACGGAATCATCCTATTCAT[C/T]TCCAAGCCACAGAAGGATGTATCAG
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600130 | ||||||||||||||||||||||||||||||||
Literature Links: |
APOBEC1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
APOBEC1 - apolipoprotein B mRNA editing enzyme catalytic subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304566.1 | 731 | Missense Mutation | AAA,AGA | K,R 236 | NP_001291495.1 | |
NM_001644.4 | 731 | Missense Mutation | AAA,AGA | K,R 236 | NP_001635.2 | |
NM_005889.3 | 731 | Missense Mutation | AAA,AGA | K,R 191 | NP_005880.2 |
Set Membership: |
HapMap |