Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGAAGAGAAAGGAAAGTGGGTGCC[A/G]GTGTCCCACGCACTGTGGACCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607211 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARD14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CARD14 - caspase recruitment domain family member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257970.1 | Intron | NP_001244899.1 | ||||
NM_024110.4 | Intron | NP_077015.2 | ||||
NM_052819.2 | Intron | NP_438170.1 | ||||
XM_011525212.1 | Intron | XP_011523514.1 | ||||
XM_011525213.1 | Intron | XP_011523515.1 | ||||
XM_011525215.1 | Intron | XP_011523517.1 | ||||
XM_011525216.1 | Intron | XP_011523518.1 | ||||
XM_011525217.1 | Intron | XP_011523519.1 | ||||
XM_011525218.2 | Intron | XP_011523520.1 | ||||
XM_017025057.1 | Intron | XP_016880546.1 | ||||
XM_017025058.1 | Intron | XP_016880547.1 |